Fmoc-Dap(Boc)-OH
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Fmoc-(N-beta-boc)-L-alpha,beta-diaminopropionic acid, 3-(tert-Butoxycarbonyl)amino-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-alanine, (S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic acid, Nα-Fmoc-Nβ-Boc-L-2,3-diaminopropionic acid, (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 3-(Boc-amino)-N-Fmoc-L-alanine
5110
AATB5110EA
460
EUR
AATB5110
Fmoc-Dap(Boc)-OH
Fmoc-Dap(Boc)-OH
FMOC-Dap(Boc)-OH is a common peptide building block.
Formel:
C₂₃H₂₆N₂O₆ Molecular Weight: 426,47 g/mol |
MDL:
MFCD00235896 CAS-Nummer: 162558-25-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...