FMOC-Asp(DABCYL)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5204
AATB5204EA 460 EUR
AATB5204
FMOC-Asp(DABCYL)-OH zur biomolekularen Markierung
FMOC-Asp(DABCYL)-OH
A building block for in-sequence Asp labeling by DABCYL.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afe81f8e811b63020cdae91513889460b78f7a22&vid=2c374ae3fbce01a4f668ba9d451f08c1355a9603&prid=web&clid=SAPDAM