FMOC-Asp(TQ2)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5206
AATB5206EA 614 EUR
AATB5206
FMOC-Asp(TQ2)-OH zur biomolekularen Markierung
FMOC-Asp(TQ2)-OH
FMOC-Asp(TQ2)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 2 (TQ2).
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b81f4cc3dee75f2669e3b355556937c559457cb&vid=b16548c971c6c35e1f57d842d2dcd876bfba9d30&prid=web&clid=SAPDAM