FMOC-Asp(TQ3)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5208
AATB5208EA 614 EUR
AATB5208
FMOC-Asp(TQ3)-OH zur biomolekularen Markierung
FMOC-Asp(TQ3)-OH
FMOC-Asp(TQ3)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 3 (TQ3).
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50386d45875d36d4040ebd23480add20a8c52429&vid=4fef9ef35399d2cf9c197c9e40ce59a47e239af6&prid=web&clid=SAPDAM