FMOC-Glu(DABCYL)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5234
AATB5234EA 460 EUR
AATB5234
FMOC-Glu(DABCYL)-OH zur biomolekularen Markierung
FMOC-Glu(DABCYL)-OH
FMOC-Glu(DABCYL)-OH is a building block for in-sequence Glu labeling by DABCYL.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6411df3f564bac207b20bff2b9d9917df2d7cb1a&vid=b0f0d55df98314a066e1d9048052bfb8f9400f63&prid=web&clid=SAPDAM