FMOC-Glu(DABCYL)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5234
AATB5234EA
460
EUR
AATB5234
FMOC-Glu(DABCYL)-OH zur biomolekularen Markierung
FMOC-Glu(DABCYL)-OH
FMOC-Glu(DABCYL)-OH is a building block for in-sequence Glu labeling by DABCYL.
- FRET building block
- Multiple applications
ADR:
5,II |