FMOC-Glu(TQ2)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5236
AATB5236EA 614 EUR
AATB5236
FMOC-Glu(TQ2)-OH zur biomolekularen Markierung
FMOC-Glu(TQ2)-OH
FMOC-Glu(TQ2)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 2 (TQ2).
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b186f5bf7c3059a70165bccbe0d0603f6f9b245d&vid=a1c59a94f099889d5b47118b35cbacc28c0476f7&prid=web&clid=SAPDAM