FMOC-Glu(TQ3)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5238
AATB5238EA 614 EUR
AATB5238
FMOC-Glu(TQ3)-OH zur biomolekularen Markierung
FMOC-Glu(TQ3)-OH
FMOC-Glu(TQ3)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 3 (TQ3).
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e68446f3a42272866df8973e65e5b53e7b078bc&vid=05d7f424cfac1d6b50d7da8f962cbdafbafa3812&prid=web&clid=SAPDAM