FMOC-Glu(TQ3)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5238
AATB5238EA
614
EUR
AATB5238
FMOC-Glu(TQ3)-OH zur biomolekularen Markierung
FMOC-Glu(TQ3)-OH
FMOC-Glu(TQ3)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 3 (TQ3).
- FRET building block
- Multiple applications
ADR:
5,II |