FMOC-Lys(TQ2)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5256
AATB5256EA
460
EUR
AATB5256
FMOC-Lys(TQ2)-OH zur biomolekularen Markierung
FMOC-Lys(TQ2)-OH
FMOC-Lys(TQ2)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 2 (TQ2).
- FRET building block
- Multiple applications
ADR:
5,II |