FMOC-Lys(TQ2)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5256
AATB5256EA 460 EUR
AATB5256
FMOC-Lys(TQ2)-OH zur biomolekularen Markierung
FMOC-Lys(TQ2)-OH
FMOC-Lys(TQ2)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 2 (TQ2).
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f47ba332eda9ec80f47a105ba411b4d0820f7e2c&vid=3eb339007d4e355b6d12c12b8f100a18c4ac9d08&prid=web&clid=SAPDAM