FMOC-Lys(TQ3)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5258
AATB5258EA 460 EUR
AATB5258
FMOC-Lys(TQ3)-OH zur biomolekularen Markierung
FMOC-Lys(TQ3)-OH
FMOC-Lys(TQ3)-OH is a building block for in-sequence Lys labeling by Tide Quencher™ 3 (TQ3).
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5094391108034f7eae0fbe9a49f6100d9a8648c&vid=061397dcae000c2836d43964554abf8b2dd38f1d&prid=web&clid=SAPDAM