7-Amino-4-methylcumarin ≥90%

Lieferant: AAT Bioquest
Achtung

51 53 52
AATB51EA 120 EUR
AATB51 AATB53 AATB52
7-Amino-4-methylcumarin ≥90%
7-Amino-4-methylcumarin

This compound is the precursor for a variety of AMC enzyme substrates.

Formel: C₁₀H₉NO₂
MDL: MFCD00006868
CAS-Nummer: 26093-31-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7232a90dc4e2e8ed29c78554b52f5d0d649282ae&vid=aae157790e6d58d2607c526391a8df661c973e39&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR