Phalloidin ≥95%

Lieferant: AAT BIOQUEST
Achtung

5301
AATB5301EA 307 EUR
AATB5301
Phalloidin ≥95%
Phalloidin

Phalloidin, a bicyclic heptapeptide toxin, binds specifically at the interface between F-actin subunits, locking adjacent subunits together.

Formel: C₃₅H₄₈N₈O₁₁S
Molecular Weight: 788,9 g/mol
CAS-Nummer: 17466-45-4
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR