Phalloidin-PEG4-DBCO zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5303
AATB5303EA
439
EUR
AATB5303
Phalloidin-PEG4-DBCO zur biomolekularen Markierung
Phalloidin-PEG4-DBCO
Phalloidin, a bicyclic heptapeptide toxin, binds specifically at the interface between F-actin subunits, locking adjacent subunits together.
- Bright signal
- High resolution imaging
ADR:
5,II |