Phalloidin-PEG4-DBCO zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5303
AATB5303EA 439 EUR
AATB5303
Phalloidin-PEG4-DBCO zur biomolekularen Markierung
Phalloidin-PEG4-DBCO
Phalloidin, a bicyclic heptapeptide toxin, binds specifically at the interface between F-actin subunits, locking adjacent subunits together.
  • Bright signal
  • High resolution imaging
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=132395994ea9562cf3deea25f4c7b26cd45394c5&vid=f6be03f8f87365d1b551a2a535226be2e09e3566&prid=web&clid=SAPDAM