Buccutite™ FOL, NHS ester (2 µmoles)
Lieferant: AAT BIOQUEST
5350
AATB5350EA 307 EUR
AATB5350
Buccutite™ FOL, NHS ester (2 µmoles)
Proteinkonjugate und Markierungen
Our Buccutite™crosslinking technology provides the most convenient and effective crosslinking method to link two biomolecules with a high conjugation yield.
  • High conjugation yield
  • Stable crosslinking
  • Minimal hands-on time
  • Robust protocol