Buccutite™ Rapid Crosslinked APC Antibody Labelling Kit
Lieferant: AAT BIOQUEST
5406
AATB5406EA
1980
EUR
AATB5406
Buccutite™ Rapid Crosslinked APC Antibody Labelling Kit
Proteinkonjugate und Markierungen
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid crosslinked APC antibody labelling kits, designed for large-scale production, offer a convenient and efficient method to label antibodies with APC, PE and iFluor® tandem dyes.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.