Buccutite™ Rapid PerCP Antibody Labelling Kit
Lieferant: AAT BIOQUEST
5407
AATB5407EA 1980 EUR
AATB5407
Buccutite™ Rapid PerCP Antibody Labelling Kit
Proteinkonjugate und Markierungen
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid PerCP antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PerCP, APC, PE and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.