Buccutite™ Rapid APC-Cy5.5 Tandem Antibody Labelling Kit
Lieferant: AAT BIOQUEST
5414
AATB5414EA
2390
EUR
AATB5414
Buccutite™ Rapid APC-Cy5.5 Tandem Antibody Labelling Kit
Proteinkonjugate und Markierungen
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid APC-Cy5.5 tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.