Buccutite™ Rapid APC-Cy7 Tandem Antibody Labelling Kit
Lieferant: AAT BIOQUEST
5415
AATB5415EA 2390 EUR
AATB5415
Buccutite™ Rapid APC-Cy7 Tandem Antibody Labelling Kit
Proteinkonjugate und Markierungen
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ rapid APC-Cy7 tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.