ReadiPrep™ Protein G-Agarose Resin
Lieferant: AAT Bioquest
55016 55017
AATB55016EA 133 EUR
AATB55016 AATB55017
ReadiPrep™ Protein G-Agarose Resin
Protein-Aufbereitungs Protein Purification Resins
ReadiPrep™ Protein G-Agarose Resin is a high-quality affinity resin designed for the isolation and purification of immunoglobulin classes, subclasses, and fragments from mammalian serum and biological fluids.
  • Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.