ReadiPrep™ Protein G-Agarose Resin
Lieferant: AAT Bioquest
ReadiPrep™ Protein G-Agarose Resin is a high-quality affinity resin designed for the isolation and purification of immunoglobulin classes, subclasses, and fragments from mammalian serum and biological fluids.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.