7-Hydroxycumarin-3-carbonsäure-N-succinimidylester ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 2,5-Dioxopyrrolidin-1-yl 7-hydroxy-2-oxo-2H-chromene-3-carboxylate

551
AATB551EA 152 EUR
AATB551
7-Hydroxycumarin-3-carbonsäure-N-succinimidylester ≥90%
7-Hydroxycumarin-3-carbonsäure-N-succinimidylester

7-Hydroxycoumarin-3-carboxylic acid, SE is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid.

Formel: C₁₄H₉NO₇
Molecular Weight: 303,23 g/mol
MDL: MFCD00037568
CAS-Nummer: 134471-24-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR