Amplite® Colorimetric Biotin Quantitation Kit
Lieferant: AAT BIOQUEST
5522
AATB5522EA 307 EUR
AATB5522
Amplite® Colorimetric Biotin Quantitation Kit
Tests Enzymtests
The avidin/streptavidin-biotin interaction is the strongest known non-covalent biological interaction (Kd = 10 to 15 M⁻¹) between a protein and its ligand.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.