Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence*
Lieferant: AAT BIOQUEST
5523
AATB5523EA
307
EUR
AATB5523
Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence*
Tests
Enzymtests
A variety of crosslinking reagents with a maleimide group are widely used for crosslinking proteins to proteins or proteins to other biomolecules.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.