Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence*
Lieferant: AAT BIOQUEST
5523
AATB5523EA 307 EUR
AATB5523
Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence*
Tests Enzymtests
A variety of crosslinking reagents with a maleimide group are widely used for crosslinking proteins to proteins or proteins to other biomolecules.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.