Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit
Lieferant: AAT BIOQUEST
5527
AATB5527EA
309
EUR
AATB5527
Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit
Tests
Enzymtests
Designed for consistent and dependable results. The Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit facilitates the quantification of maleimide groups on maleimide-activated APC proteins using a proprietary sensor, Maleimide 555™, which has an absorption peak at ~555 nm.
- Robust protocol
- This quantitation kit is compatible with various detection systems, including traditional cuvettes, NanoDrop™ Spectrophotometers, and 96-well absorbance plate readers
- Six-month shelf life
Maleimide-based crosslinking reagents are commonly used for conjugating proteins to other proteins or biomolecules.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.