7-Hydroxycoumarin-4-acetic acid succinimidyl ester ≥90%
Lieferant: AAT BIOQUEST
553
AATB553EA
152
EUR
AATB553
7-Hydroxycoumarin-4-acetic acid succinimidyl ester ≥90%
7-Hydroxycoumarin-4-acetic acid succinimidyl ester
7-Hydroxycoumarin-4-acetic acid is used for labeling peptides and proteins to give blue fluorescence.
- Multiple Bioconjugation Applications
- 90% Purity
Bitte beachten For research use only.