7-Hydroxy-4-methylcoumarin-3-acetic acid ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
4-Methylumbelliferone-3-acetic Acid, 2-(7-Hydroxy-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid
554
AATB554EA
120
EUR
AATB554
7-Hydroxy-4-methylcoumarin-3-acetic acid ≥95%
7-Hydroxy-4-methylcoumarin-3-acetic acid
7-Hydroxy-4-methylcoumarin-3-acetic acid is a blue fluorophore that has pH-dependent and environment-sensitive fluorescence.
Formel:
C₁₂H₁₀O₅ Molecular Weight: 234,21 g/mol |
MDL:
MFCD00467589 CAS-Nummer: 5852-10-8 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...