7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester

Lieferant: AAT BIOQUEST

Synonyme: N-Succinimidyl 7-Hydroxy-4-methylcoumarin-3-acetate, N-Succinimidyl 4-Methylumbelliferone-3-acetate, 2,5-Dioxopyrrolidin-1-yl 2-(7-hydroxy-4-methyl-2-oxo-2H-chromen-3-yl)acetate, 7-Hydroxy-4-methylcoumarin-3-acetic acid N-Succinimidyl ester, 4-Methylumbelliferone-3-acetic acid N-Succinimidyl ester

556
AATB556EA 152 EUR
AATB556
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester

7-Hydroxy-4-methylcoumarin-3-acetic acid, SE is an amine-reactive blue fluorophore that has pH-dependent and environment-sensitive fluorescence.


  • Multiple Bioconjugation Applications
  • pH-independent Fluorescence


Bitte beachten For research use only.

Formel: C₁₆H₁₃NO₇
Molecular Weight: 331,28 g/mol
MDL: MFCD00467590
CAS-Nummer: 96735-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR