7-Methoxycoumarin-4-acetic acid ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 7-Methoxy-2-oxo-2H-1-benzopyran-4-acetic acid, Mca, 2-(7-Methoxy-2-oxo-2H-chromen-4-yl)acetic acid, (7-Methoxycoumarin-4-yl)acetic acid

557
AATB557EA 307 EUR
AATB557
7-Methoxycoumarin-4-acetic acid ≥90%
7-Methoxycoumarin-4-acetic acid

7-Methoxycoumarin-4-acetic acid (MCA) is widely used for developing FRET peptide substrates for analyzing protease activities.

Formel: C₁₂H₁₀O₅
Molecular Weight: 234,207 g/mol
MDL: MFCD00009774
CAS-Nummer: 62935-72-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR