7-Methoxy-4-coumarinylacetic acid N-succinimidyl ester ≥90%

Lieferant: AAT BIOQUEST

Synonyme: MCA succinimidyl ester, 7-Methoxycoumarin-4-acetic acid succinimidyl ester

558
AATB558EA 230 EUR
AATB558
7-Methoxy-4-coumarinylacetic acid N-succinimidyl ester ≥90%
7-Methoxy-4-coumarinylacetic acid N-succinimidyl ester

7-Methoxycoumarin-4-acetic acid (MCA) is widely used for developing FRET peptide substrates for analyzing protease activities.


  • Multiple Bioconjugation Applications
  • 90% Purity


Bitte beachten For research use only.

Formel: C₁₆H₁₃NO₇
Molecular Weight: 331,28 g/mol
CAS-Nummer: 359436-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR