cBSA *Optimised for Maximum Immunisation Response* (10 mg)
Lieferant: AAT BIOQUEST
5600
AATB5600EA 307 EUR
AATB5600
cBSA *Optimised for Maximum Immunisation Response* (10 mg)
Seren
cBSA is the bovine serum albumin modified with ethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).
  • Lyophilized powder
  • Excellent quality control
  • Stronger immune response
  • 60000 molecular weight
Bitte beachten For research use only.