ReadiPrep™ Nickel NTA Magnetic Beads
Lieferant: AAT BIOQUEST
56000 56001
AATB56000EA 130 EUR
AATB56000 AATB56001
ReadiPrep™ Nickel NTA Magnetic Beads
Protein-Aufbereitungs Proteinreinigungs-Reagenzien
ReadiPrep™ Nickel NTA magnetic beads provide a fast and simple solution for small-scale affinity purification of histidine-tagged proteins.
  • Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.