N-Succinimidyl 7-Methoxycoumarin-3-carboxylate ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 7-methoxy-2-oxo-2H-chromene-3-carboxylate

563
AATB563EA 152 EUR
AATB563
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate ≥90%
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate

7-Methoxycoumarin-3-carboxylic acid, succinimidyl ester is an excellent amino-reactive tag that has strong blue fluorescence.

Formel: C₁₅H₁₁NO₇
Molecular Weight: 317,25 g/mol
MDL: MFCD00467593
CAS-Nummer: 150321-92-9
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR