3-Carboxy-6,8-difluoro-7-hydroxycoumarin ≥90% [equivalent to Pacific Blue succinimidyl ester]

Lieferant: AAT BIOQUEST

Synonyme: PacBlue succinimidyl ester, NHS Pacific Blue succinimidyl ester

570
AATB570EA 307 EUR
AATB570
3-Carboxy-6,8-difluoro-7-hydroxycoumarin ≥90% [equivalent to Pacific Blue succinimidyl ester]
3-Carboxy-6,8-difluoro-7-hydroxycoumarin

PacBlue succinimidyl ester is the same molecule to Pacific Blue™ succinimidyl ester (Pacific Blue™ is the trademark of ThermoFisher).


  • Multiple Bioconjugation Applications
  • 90% Purity


Bitte beachten For research use only.

Formel: C₁₄H₇F₂NO₇
Molecular Weight: 339,2 g/mol
CAS-Nummer: 215868-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR