ReadiLink™ Rapid XFD594 Antibody Labelling Kit
Lieferant: AAT BIOQUEST
5736
AATB5736EA
687
EUR
AATB5736
ReadiLink™ Rapid XFD594 Antibody Labelling Kit
Proteinkonjugate und Markierungen
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with XFD dyes (equivalent to Alexa Fluor®).
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Lieferumfang: Shipped at RT.
Bitte beachten Research use only.