Basic blue 12 einfaches Isomer DNA-Farbstoff

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Nile Blue A chloride, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium chloride

59
AATB59EA 218 EUR
AATB59
Basic blue 12 einfaches Isomer DNA-Farbstoff
Nile blue chloride (Basic blue 12)

Nile blue A is a stain used in biology and histology.


  • Does not require UV trans-illumination

Formel: C₂₀H₂₀ClN₃O
Lagertemperatur: Gefrierschrank
MDL: MFCD00011936
CAS-Nummer: 2381-85-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8600627af9b05e3d58fb630a267e288b6f2654ae&vid=727a5a7073445cf4af153d422ff7b935d82a8c4c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR