Nile blue A ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Nile blue A sulphate, Nile blue sulphate, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium sulphate, Basic Blue 12
60
AATB60EA
120
EUR
AATB60
Nile blue A ≥95%
Nile blue A
Nile Blue A is certified for use in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.
Formel:
C₄₀H₄₀N₆O₆S |
MDL:
MFCD00064529 CAS-Nummer: 3625-57-8 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...