Amplite® Colorimetric Endotoxin Detection Kit
Lieferant: AAT BIOQUEST
60007
AATB60007EA
920
EUR
AATB60007
Amplite® Colorimetric Endotoxin Detection Kit
Tests
Enzymtests
Lipopolysaccharide (LPS), also known as endotoxin, is the major component of the outer membranes of Gram-negative bacteria.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.