Amplite® Colorimetric Endotoxin Detection Kit
Lieferant: AAT BIOQUEST
60007
AATB60007EA 920 EUR
AATB60007
Amplite® Colorimetric Endotoxin Detection Kit
Tests Enzymtests
Lipopolysaccharide (LPS), also known as endotoxin, is the major component of the outer membranes of Gram-negative bacteria.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Bitte beachten For research use only.