Amino-Modifier C6 dT

Lieferant: AAT BIOQUEST

6002
AATB6002EA 206 EUR
AATB6002
Amino-Modifier C6 dT
Amino-Modifier C6 dT

Amino-Modifier C6 dT is a convenient phosphoramidite reagent used to introduce an amino group from dT in oligo synthesis.


  • Stable for 6 months


Quality tested in-house

Molecular Weight: 995.05 g/mol
CAS-Nummer: 178925-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR