3'-DABCYL CPG 1000 Å zur Oligonukleotidmarkierung
Lieferant: AAT BIOQUEST
|
Achtung
|
6008
AATB6008EA
1170
EUR
AATB6008
3'-DABCYL CPG 1000 Å zur Oligonukleotidmarkierung
3'-DABCYL CPG
DABCYL is one of the most commonly used dark quenchers for labeling oligos and peptides.
- FRET building block
- Multiple applications
ADR:
5,II |