3'-DABCYL CPG 1000 Å zur Oligonukleotidmarkierung
Lieferant: AAT BIOQUEST
Achtung
6008
AATB6008EA 1170 EUR
AATB6008
3'-DABCYL CPG 1000 Å zur Oligonukleotidmarkierung
3'-DABCYL CPG
DABCYL is one of the most commonly used dark quenchers for labeling oligos and peptides.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0868006ad2901230ceb6f09cd2bcd49f057776ab&vid=72f3ed9e59559888db15f1bbfb9608873b048aaa&prid=web&clid=SAPDAM