5'-DABCYL C6 phosphoramidite zur Oligonukleotidmarkierung
Lieferant: AAT BIOQUEST
Achtung
6009
AATB6009EA 1480 EUR
AATB6009
5'-DABCYL C6 phosphoramidite zur Oligonukleotidmarkierung
5'-DABCYL C6 phosphoramidite
DABCYL is one of the most commonly used dark quenchers for labeling oligos and peptides.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=711cc488f79307090c44314329d2bfb9d95d83c2&vid=8082326d39b4aae2564b93f52329f89c669d7864&prid=web&clid=SAPDAM