ReadiUse™ Spin Filter (5), 10KD
Lieferant: AAT BIOQUEST
60502
AATB60502EA 113 EUR
AATB60502
ReadiUse™ Spin Filter (5), 10KD
Filter Zentrifugaleinheiten
ReadiUse™ 10 Kd spin filter is a disposable ultrafiltration device for the concentration of biological samples such as antibodies.
  • Compatible with both swing bucket or fixed angle instruments
  • Designed to prevent membrane fouling
  • Low internal surface area
  • High material recovery from dilute solutions
Bitte beachten For research use only.