6-TAMRA CPG 1000 Å zur Oligonukleotidmarkierung
Lieferant: AAT BIOQUEST
Achtung
6051
AATB6051EA 769 EUR
AATB6051
6-TAMRA CPG 1000 Å zur Oligonukleotidmarkierung
6-TAMRA CPG
The light-absorbing properties of TAMRA, and spectral overlap with several commonly used fluorophores - including FAM, HEX, TET and JOE, make it useful as a FRET acceptor for the dual labeled FRET probes such as molecular beacons.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14291d318c661e8d14d1af94220cdb8272484768&vid=6464019037ce42a9f9d0fa5b99a6c99cbcfcdfc3&prid=web&clid=SAPDAM