6-TAMRA CPG 1000 Å zur Oligonukleotidmarkierung
Lieferant: AAT BIOQUEST
|
Achtung
|
6051
AATB6051EA
769
EUR
AATB6051
6-TAMRA CPG 1000 Å zur Oligonukleotidmarkierung
6-TAMRA CPG
The light-absorbing properties of TAMRA, and spectral overlap with several commonly used fluorophores - including FAM, HEX, TET and JOE, make it useful as a FRET acceptor for the dual labeled FRET probes such as molecular beacons.
- FRET building block
- Multiple applications
ADR:
5,II |