Cy3 phosphoramidite

Lieferant: AAT BIOQUEST

6070
AATB6070EA 282 EUR
AATB6070
Cy3 phosphoramidite
Cy3 phosphoramidite

Two cyanine derivatives, Cy3 and Cy5 phosphoramidites, are the most popular fluorescent labeling reagents for preparing fluorescence-labeled oligonucleotides.


  • Stable for 6 months


Quality tested in-house

Molecular Weight: 953,63 g/mol
CAS-Nummer: 182873-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR