Cy5 phosphoramidite
Lieferant: AAT BIOQUEST
6071
AATB6071EA
282
EUR
AATB6071
Cy5 phosphoramidite
Cy5 phosphoramidite
Two cyanine derivatives, Cy3 and Cy5 phosphoramidites, are the most popular fluorescent labeling reagents for preparing fluorescence-labeled oligonucleotides.
- Stable for 6 months
Quality tested in-house
Bitte beachten For Research Use Only