Pyronin G ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Pyronin Y, 3,6-Bis(dimethylamino)xanthyliumchlorid

61
AATB61EA 120 EUR
AATB61
Pyronin G ≥95%
Pyronin G

Pyronin Y is widely used in combination with methyl green to selectively and differentially stain nucleic acids following polyacrylamide gel electrophoresis.

Formel: C₁₇H₁₉ClN₂O
Molecular Weight: 302,8 g/mol
MDL: MFCD00011725
CAS-Nummer: 92-32-0
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR