5-(2-Aminoethylamino)-1-naphthalenesulphonic acid ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 5-((2-Aminoethyl)amino)naphthalene-1-sulphonic acid, EDANS, acid

610
AATB610EA 152 EUR
AATB610
5-(2-Aminoethylamino)-1-naphthalenesulphonic acid ≥95%
5-(2-Aminoethylamino)-1-naphthalenesulphonic acid

EDANS is one of the most popular donors for developing FRET-based nucleic acid probes (such as molecular beacons) and protease substrates.

Formel: C₁₂H₁₄N₂O₃S
Molecular Weight: 266,315 g/mol
MDL: MFCD00066932
CAS-Nummer: 50402-56-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR