EDANS C5 maleimide fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
619
AATB619EA
143
EUR
AATB619
EDANS C5 maleimide fluoreszierende Probe
EDANS C5 maleimide
EDANS is one of the most popular donors for developing FRET-based nucleic acid probes (such as Molecular Beacons) and protease substrates.
- Improved water solubility
- Easily purified
- Strong fluorescence
Molecular Weight:
417,44 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...