8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: APTS

624
AATB624EA 120 EUR
AATB624
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt ≥95%
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt

This water-soluble reagent has strong green fluorescence with great Stokes shift.

Formel: C₁₆H₈NNa₃O₉S₃
Molecular Weight: 523,40 g/mol
MDL: MFCD00674494
CAS-Nummer: 196504-57-1
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR