Resorufin, sodium salt ≥90%

Lieferant: AAT BIOQUEST
Achtung

65
AATB65EA 120 EUR
AATB65
Resorufin, sodium salt ≥90%
Resorufin, sodium salt

Resorufin is used as a calibration standard for resorufin-derived enzyme substrates.

Formel: C₁₂H₆NNaO₃
Molecular Weight: 235,17 g/mol
MDL: MFCD00005037
CAS-Nummer: 34994-50-8
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR