576/589 NHS Ester, Bodi Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Py-BODIPY-NHS ester, 2,5-Dioxopyrrolidin-1-yl 3-(5,5-difluoro-7-(1H-pyrrol-2-yl)-5H-5l4,6l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-3-yl)propanoate
661
AATB661EA
439
EUR
AATB661
576/589 NHS Ester, Bodi Fluor™ fluoreszierende Probe
576/589 NHS Ester
Bodi Fluor™ 576/589 is chemically the same molecule to BODIPY™ 576/589 dye.
- Bright signal
- Highly cell permeable
- High extinction coefficient
- Insensitive to polarity/pH
- High fluorescence quantum yield
Molecular Weight:
426,19 g/mol |
MDL:
MFCD32858277 CAS-Nummer: 201998-61-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...