NED Dye qPCR Calibration Solution *10000X*
Lieferant: AAT BIOQUEST
67031
AATB67031EA 420 EUR
AATB67031
NED Dye qPCR Calibration Solution *10000X*
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
NED dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
  • Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Bitte beachten For research use only.