FAM Dye qPCR Calibration Solution *10000X*
Lieferant: AAT BIOQUEST
67032
AATB67032EA 420 EUR
AATB67032
FAM Dye qPCR Calibration Solution *10000X*
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
FAM dye qPCR calibration solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
  • Stable for 12 months
Calibration and verification should be run every six months or as needed.
Bitte beachten For research use only.